Skip to Content
Merck
All Photos(1)

Key Documents

36011

Sigma-Aldrich

Picric acid solution

0.9-1.1% (alkalimetric)

Synonym(s):

2,4,6-Trinitrophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)3C6H2OH
CAS Number:
Molecular Weight:
229.10
Beilstein:
423400
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

form

aqueous solution

shelf life

5 yr

concentration

0.9-1.1% (alkalimetric)

density

0.99-1.01 g/cm3

application(s)

hematology
histology

storage temp.

room temp

SMILES string

Oc1c(cc(cc1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H3N3O7/c10-6-4(8(13)14)1-3(7(11)12)2-5(6)9(15)16/h1-2,10H

InChI key

OXNIZHLAWKMVMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

Precipitation eventually caused by low temperature may be dissolved simply by a waterbath at ~40°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dustin Staloch et al.
Journal of molecular medicine (Berlin, Germany), 93(10), 1085-1093 (2015-07-05)
The current study aims to identify the pro-fibrogenic role of Gremlin, an endogenous antagonist of bone morphogenetic proteins (BMPs) in chronic pancreatitis (CP). CP is a highly debilitating disease characterized by progressive pancreatic inflammation and fibrosis that ultimately leads to
Alex S Borges et al.
Inorganic chemistry, 51(23), 12867-12878 (2012-11-16)
Six new lanthanide complexes of stoichiometric formula (C)(2)[Ln(Pic)(5)]--where (C) is a imidazolium cation coming from the ionic liquids 1-butyl-3-methylimidazolium picrate (BMIm-Pic), 1-butyl-3-ethylimidazolium picrate (BEIm-Pic), and 1,3-dibutylimidazolium picrate (BBIm-Pic), and Ln is Eu(III) or Gd(III) ions--have been prepared and characterized. To
D Toki et al.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons, 14(9), 2126-2136 (2014-10-14)
The aim of this study was to investigate the role of infiltrating macrophages in renal allograft fibrosis. Forty-six protocol renal allograft biopsies obtained 1 year after transplantation were stained with Sirius red to quantify fibrosis and double stained with CD68
Dmitry Momotenko et al.
Analytical chemistry, 84(15), 6630-6637 (2012-07-14)
This manuscript presents a push-pull electrochemical scanner able to image reactivity of initially dry surfaces by scanning electrochemical microscopy (SECM) and to probe molecules present or generated at the surface by mass spectrometry (MS). The proof-of-concept is demonstrated by coupling
Debby Kruijsen et al.
Journal of virology, 87(13), 7550-7557 (2013-05-03)
Infants are protected from a severe respiratory syncytial virus (RSV) infection in the first months of life by maternal antibodies or by prophylactically administered neutralizing antibodies. Efforts are under way to produce RSV-specific antibodies with increased neutralizing capacity compared to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service