Skip to Content
Merck
All Photos(1)

Key Documents

35811

Supelco

2,4-Dichlorophenol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3OH
CAS Number:
Molecular Weight:
163.00
Beilstein:
742467
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
39050432
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

209-210 °C (lit.)

mp

42-43 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Oc1ccc(Cl)cc1Cl

InChI

1S/C6H4Cl2O/c7-4-1-2-6(9)5(8)3-4/h1-3,9H

InChI key

HFZWRUODUSTPEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Dichlorophenol (2,4 DCP) is a toxic chlorophenol belonging to the class of volatile organic compound (VOC) mainly used as pesticide, herbicide, wood, leather and glue preservatives.[1]

Application

2,4-Dichlorophenol may be used as an analytical reference standard for the determination of the analyte in water[2] and soil[3] samples by various chromatography methods.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

231.1 °F - closed cup

Flash Point(C)

110.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Determination of 2, 4-dichlorophenoxyacetic acid and its major transformation product in soil samples by liquid chromatographic analysis.
de Amarante Jr OP, et al.
Talanta, 60(1), 115-121 (2003)
Dispersive liquid--liquid microextraction based on solidification of floating organic droplet followed by high-performance liquid chromatography with ultraviolet detection and liquid chromatography-tandem mass spectrometry for the determination of triclosan and 2, 4-dichlorophenol in water samples
Zheng C, et al.
Journal of Chromatography A, 1218(25), 3830-3836 (2011)
Removal of 2, 4-dichlorophenol from wasterwater by vacuum membrane distillation using hydrophobic PPESK hollow hiber membrane.
Jin Z, et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 18(12), 1543-1547 (2007)
Santiago Gómez-Quero et al.
Nanotechnology, 23(29), 294002-294002 (2012-06-30)
Catalytic hydrodechlorination (HDC) is an effective means of detoxifying chlorinated waste. Gold nanoparticles supported on Fe(3)O(4) have been tested in the gas phase (1 atm, 423 K) HDC of 2,4-dichlorophenol. Two 1% w/w supported gold catalysts have been prepared by:
Jiang Xu et al.
Journal of hazardous materials, 244-245, 628-636 (2012-11-28)
The Pd/FeFe(3)O(4) nanocomposites showed higher dechlorination efficiency of 2,4-dichlorophenol (2,4-DCP) rather than bare Pd/Fe nanoparticles in the batch dechlorination experiments. nFe(3)O(4) provided a convenient way to recycle the nanocomposites with an external magnetic field and significantly enhanced 2,4-DCP dechlorination. 2.0

Protocols

Analytical standard separation of various phenols for research and industrial applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service