Skip to Content
Merck
All Photos(1)

Key Documents

35516

Supelco

Patulin-13C7 solution

~25 μg/mL in acetonitrile, analytical standard

Synonym(s):

4-Hydroxy-4H-furo[3,2-c]pyran-2(6H)-one-13C7

Sign Into View Organizational & Contract Pricing

Select a Size

1.2 ML
CHF 2’320.00

CHF 2’320.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
1.2 ML
CHF 2’320.00

About This Item

Empirical Formula (Hill Notation):
13C7H6O4
CAS Number:
Molecular Weight:
161.07
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

CHF 2’320.00


Available to ship on31 March 2025Details


Request a Bulk Order

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

~25 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

mass shift

M+7

storage temp.

−20°C

SMILES string

O[13CH]1O[13CH2][13CH]=[13C]2O[13C](=O)[13CH]=[13C]12

InChI

1S/C7H6O4/c8-6-3-4-5(11-6)1-2-10-7(4)9/h1,3,7,9H,2H2/i1+1,2+1,3+1,4+1,5+1,6+1,7+1

InChI key

ZRWPUFFVAOMMNM-BNUYUSEDSA-N

General description

Patulin-13C7 is the 13C isotope-labelled analog of mycotoxin, Patulin

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥95.5% (HPLC)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service