Skip to Content
Merck
All Photos(1)

Key Documents

34854-M

Sigma-Aldrich

Chloroform

suitable for HPLC, ≥99.8%, amylene stabilized

Synonym(s):

Methylidyne trichloride, Trichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein:
1731042
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
grade:
HPLC grade
Assay:
≥99.8%
technique(s):
HPLC: suitable
application(s):
general analytical
bp:
60.5-61.5 °C (lit.)
vapor pressure:
160 mmHg ( 20 °C)

grade

HPLC grade

vapor density

4.1 (vs air)

vapor pressure

160 mmHg ( 20 °C)

Assay

≥99.8%

form

liquid

quality

amylene stabilized

technique(s)

HPLC: suitable

impurities

Free acid, in accordance
acetone, aldehyde, in accordance (nessler test)
free chlorine (Cl), in accordance
≤0.0005% non-volatile matter
≤0.001% free acid (as HCl)
<0.01% water (Karl Fischer)

evapn. residue

≤0.0005%

color

APHA: ≤10

transmittance

250 nm, ≥50%
260 nm, ≥85%
280 nm, ≥99%

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

solubility

water: soluble (8.7 g/l at 23 °C (73 °F) - OECD Test Guideline 105)

density

1.48 g/mL at 25 °C
1.492 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): in accordance

cation traces

Pb: ≤0.05 mg/kg

absorption

in accordance

λ

H2O reference

UV absorption

λ: 250 nm Amax: ≤1.00
λ: 260 nm Amax: ≤0.30
λ: 280 nm Amax: ≤0.005

suitability

passes test for reaction against H2SO4
passes test for suitability of determ. w. dithizone

application(s)

general analytical

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroform, a halogenated hydrocarbon, is widely employed as a solvent. Its photochemical degradation has been investigated in the presence of TiO2 aqueous suspensions (wavelength range of 310-380nm).[1]

Application

Chloroform has been employed for the ion exchange isolation of sulfatides from the urinary samples by using DEAE-cellulose membranes before the tandem mass spectrometric analysis.[2] It has been used as a solvent for the Langmuir-Blodgett (LB) deposition of lipid bilayers over mica sheets (silver coated back).†

Other Notes

Important notice
  • The article number 34854-4X2.5L-M will be discontinued. Please order the single bottle 34854-2.5L-M which is physically identical with the same exact specifications.
  • The article number 34854-6X1L-M will be discontinued. Please order the single bottle 34854-1L-M which is physically identical with the same exact specifications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ladislav Kuchař et al.
Clinica chimica acta; international journal of clinical chemistry, 425, 153-159 (2013-07-11)
Prediagnostic steps in suspected metachromatic leukodystrophy (MLD) rely on clinical chemical methods other than enzyme assays. We report a new diagnostic method which evaluates changes in the spectrum of molecular types of sulfatides (3-O-sulfogalactosyl ceramides) in MLD urine. The procedure

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service