Skip to Content
Merck
All Photos(1)

Key Documents

34572

Supelco

Thiabendazole-(benzimidazole-3a,4,5,6,7,7a-13C6)

PESTANAL®, analytical standard

Synonym(s):

2-(4-Thiazolyl)benzimidazole-3a,4,5,6,7,7a-13C6, Thiabendazole-13C6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C4H7N3S
Molecular Weight:
207.20
UNSPSC Code:
77101502
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

SMILES string

C1(C2=N[13C]3=[13CH][13CH]=[13CH][13CH]=[13C]3N2)=CSC=N1

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service