Skip to Content
Merck
All Photos(1)

Key Documents

34231

Supelco

Naftalofos

PESTANAL®, analytical standard

Synonym(s):

O,O-Diethyl N-hydroxynaphthalimide phosphate, Naphthalophos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16NO6P
CAS Number:
Molecular Weight:
349.28
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=O)(OCC)ON1C(=O)c2cccc3cccc(C1=O)c23

InChI

1S/C16H16NO6P/c1-3-21-24(20,22-4-2)23-17-15(18)12-9-5-7-11-8-6-10-13(14(11)12)16(17)19/h5-10H,3-4H2,1-2H3

InChI key

QNSIFYWAPWSAIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Naftalofos is an organophosphorus pesticide primarily used to control pests in various agricultural applications. Its mode of action involves inhibition of the acetylcholinesterase (AChE) enzyme.

Application

Naftalofos may be used as an analytical reference standard for the determination of the target analyte in food and environmental samples by square wave voltammetric detection method using novel modified reduced graphene oxide (RGO)/chitosan (CS) on a glassy carbon electrode.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M T Santamarina et al.
Parasitology research, 74(6), 581-585 (1988-01-01)
The effect of the adult worms and migrating L1 larvae of Trichinella spiralis on the production of specific IgE antibodies was determined in BCF1 mice. To achieve this, we combined the effect of two anthelminthics: thiabendazole, to produce chemosterilization of
Mixtures of anthelmintics: a strategy against resistance.
N Anderson et al.
Australian veterinary journal, 65(2), 62-64 (1988-02-01)
K M Dash
Australian veterinary journal, 63(2), 45-47 (1986-02-01)
Following the failure of anthelmintic treatment to control an outbreak of trichostrongylosis in sheep, multiple resistance to levamisole and oxfendazole was confirmed in field strains of Trichostrongylus colubriformis at the CSIRO Pastoral Research Laboratory, Armidale. Resistance in Trichostrongylus spp to
L F Le Jambre et al.
Veterinary parasitology, 128(1-2), 83-90 (2005-02-24)
The development of moxidectin resistance (MOX-R) in sheep parasitic gastrointestinal nematodes already carrying multiple resistances to other anthelmintic groups has made control of these strains very difficult. The anthelmintic resistance patterns of MOX-R strains of Trichostrongylus colubriformis and Haemonchus contortus
N Sangster
Parasitology, 113 Suppl, S201-S216 (1996-01-01)
Anthelmintic resistance has compromised the control of nematode parasites in several animal-based industries. Studies of resistance have not only improved our understanding of this phenomenon but also shed light on physiological systems of parasitic helminths. In addition, research on molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service