34167
Dihexyl phthalate-3,4,5,6-d4
analytical standard
Synonym(s):
Di-n-hexyl phthalate-d4
About This Item
Recommended Products
grade
analytical standard
Quality Level
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
cleaning products
cosmetics
environmental
food and beverages
personal care
format
neat
mass shift
M+4
SMILES string
[2H]c1c([2H])c([2H])c(C(=O)OCCCCCC)c(c1[2H])C(=O)OCCCCCC
InChI
1S/C20H30O4/c1-3-5-7-11-15-23-19(21)17-13-9-10-14-18(17)20(22)24-16-12-8-6-4-2/h9-10,13-14H,3-8,11-12,15-16H2,1-2H3/i9D,10D,13D,14D
InChI key
KCXZNSGUUQJJTR-LSQQNFJSSA-N
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
350.6 °F
Flash Point(C)
177 °C
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service