Skip to Content
Merck
All Photos(1)

Documents

33890

Supelco

PCB No 209 solution

10 μg/mL in heptane, analytical standard

Synonym(s):

Decachlorobiphenyl solution, 2,2′,3,3′,4,4′,5,5′,6,6′−PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12Cl10
CAS Number:
Molecular Weight:
498.66
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in heptane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

SMILES string

Clc1c(Cl)c(Cl)c(c(Cl)c1Cl)-c2c(Cl)c(Cl)c(Cl)c(Cl)c2Cl

InChI

1S/C12Cl10/c13-3-1(4(14)8(18)11(21)7(3)17)2-5(15)9(19)12(22)10(20)6(2)16

InChI key

ONXPZLFXDMAPRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photodegradation of decachlorobiphenyl.
M Koshioka et al.
Bulletin of environmental contamination and toxicology, 38(3), 409-415 (1987-03-01)
A morphologic and physiologic bioassay for quantifying alterations in the lung following experimental chronic inhalation of tobacco smoke.
G L Huber et al.
Bulletin europeen de physiopathologie respiratoire, 17(2), 269-327 (1981-01-01)
Perchlorination reaction applied to the rapid determination of PCBs.
K Takamiya
Bulletin of environmental contamination and toxicology, 30(5), 600-605 (1983-05-01)
Xing Han et al.
Toxicology, 255(3), 177-186 (2008-11-22)
2,2',3,3',4,4',5,5',6,6'-Decachlorobiphenyl (PCB 209) is a fully chlorinated, non-coplanar biphenyl. To demonstrate that PCB 209 is not likely to exhibit human health hazards common to coplanar PCBs it was tested for cytochrome P450 (P450) enzyme induction potentials, genetic toxicity, and endocrine-modulating
A R Shoaf et al.
Journal of bioluminescence and chemiluminescence, 11(1), 9-22 (1996-01-01)
The reactions between superoxide free radical anion (.O2-) with the halocarbons CCl4. CHCl3, BrCH2CH2Br(EDB), decachlora-biphenyl (DCBP), and 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD) in dimethyl sulphoxide (DMSO) results in the emission of chemiluminescence (CL). The chemiluminescence reactions are characterized as having biphasic second order

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service