Skip to Content
Merck
All Photos(1)

Key Documents

33270

Sigma-Aldrich

L-2,3-Diaminopropionic acid hydrochloride

≥97.0%

Synonym(s):

3-Amino-L-alanine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H8N2O2 · HCl
CAS Number:
Molecular Weight:
140.57
Beilstein:
4263944
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥97.0% (AT)
≥97.0%

form

solid

optical activity

[α]20/D +24±2°, c = 2% in 0.5 M HCl

loss

<1% loss on drying

mp

231-233 °C (dec.)

application(s)

peptide synthesis

SMILES string

O=C(O)[C@H](CN)N.Cl

InChI

1S/C3H8N2O2.ClH/c4-1-2(5)3(6)7;/h2H,1,4-5H2,(H,6,7);1H/t2-;/m0./s1

InChI key

SKWCZPYWFRTSDD-DKWTVANSSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Substrate for (Km =1mM), and competitive inhbitor (Ki =1mM, L-homoserine as substrate) of cystathionase from rat liver[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rat liver L-diaminopropionate ammonia lyase. Identification as cystathionase.
I K Mushahwar et al.
The Journal of biological chemistry, 248(21), 7407-7411 (1973-11-10)
Giovanni N Roviello et al.
Amino acids, 43(6), 2537-2543 (2012-06-13)
In this work, we report a synthetic approach to a Fmoc-protected nucleoamino acid, based on L-diaminopropanoic acid, carrying the DNA nucleobase on the alpha-amino group by means of an amide bond, suitable for the solid-phase synthesis of novel nucleopeptides of
Johnson Cheung et al.
Molecular microbiology, 74(3), 594-608 (2009-09-25)
Siderophores are iron-scavenging molecules produced by many microbes. In general, they are synthesized using either non-ribosomal peptide synthetase (NRPS) or NRPS-independent siderophore (NIS) pathways. Staphylococcus aureus produces siderophores, of which the structures of staphyloferrin A and staphyloferrin B are known.
Joana R Viola et al.
Bioorganic & medicinal chemistry letters, 22(17), 5635-5638 (2012-08-04)
Here we report a novel class of peptides-d-diaminopropionic acids (Dap)-for gene delivery. These peptides have attractive properties for gene delivery, and the advantage that they can be easily manipulated in relation to their composition, abiding with tailored-design. We characterized the
J N Kalyani et al.
Journal of bacteriology, 194(20), 5604-5612 (2012-08-21)
Diaminopropionate ammonia lyase (DAPAL) is a pyridoxal-5'phosphate (PLP)-dependent enzyme that catalyzes the conversion of diaminopropionate (DAP) to pyruvate and ammonia and plays an important role in cell metabolism. We have investigated the role of the ygeX gene of Escherichia coli

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service