Skip to Content
Merck
All Photos(1)

Key Documents

32928

Supelco

15-Acetyldeoxynivalenol

reference material

Synonym(s):

15-O-Acetyl-4-deoxynivalenol from Fusarium graminearum, 15-Acetoxy-3α,7α-dihydroxy-12,13-epoxytrichothec-9-en-8-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22O7
CAS Number:
Molecular Weight:
338.35
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
reference material

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

CC(=O)OC[C@]12[C@H](O)C(=O)C(C)=C[C@H]1O[C@@H]3[C@H](O)C[C@@]2(C)[C@]34CO4

InChI

1S/C17H22O7/c1-8-4-11-16(6-22-9(2)18,13(21)12(8)20)15(3)5-10(19)14(24-11)17(15)7-23-17/h4,10-11,13-14,19,21H,5-7H2,1-3H3/t10-,11-,13-,14-,15-,16-,17+/m1/s1

InChI key

IDGRYIRJIFKTAN-HTJQZXIKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

15-Acetyldeoxynivalenols belong to the class of type B trichothecene mycotoxins, generated by several Fusarium fungi.[1]

Application

15-Acetyldeoxynivalenol may be used as an analytical reference standard for the determination of the analyte in:
  • Pig urine and maize samples by high performance liquid chromatography-mass spectrometry (HPLC-MS).[2]
  • Milk samples by liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).[3]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of major B-trichothecenes and the de-epoxy-metabolite of deoxynivalenol in pig urine and maize using high-performance liquid chromatography-mass spectrometry.
Razzazi-Fazeli E, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 796(1), 21-33 (2003)
Determination of 12 type A and B trichothecenes in cereals by liquid chromatography- electrospray ionization tandem mass spectrometry.
Klotzel M, et al.
Journal of Agricultural and Food Chemistry, 53(23), 8904-8910 (2005)
Determination of mycotoxins in bovine milk by liquid chromatography tandem mass spectrometry.
S?rensen LK and Elb?k TH
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 820(2), 183-196 (2005)
Jing-Bo Zhang et al.
Mycological research, 111(Pt 8), 967-975 (2007-08-25)
A large number of isolates from the Fusarium graminearum clade representing all regions in China with a known history of Fusarium head blight (FHB) epidemics in wheat were assayed using PCR to ascertain their trichothecene mycotoxin chemotypes and associated phylogenetic
Nadia Ponts et al.
FEBS letters, 581(3), 443-447 (2007-01-26)
Effect of exogenous H(2)O(2) and catalase was tested in liquid cultures of the deoxynivalenol and 15-acetyldeoxynivalenol-producing fungus Fusarium graminearum. Accordingly to previous results, H(2)O(2) supplementation of the culture medium leads to increased toxin production. This study indicates that this event

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service