Skip to Content
Merck
All Photos(2)

Key Documents

32919

Supelco

Imazosulfuron

PESTANAL®, analytical standard

Synonym(s):

1-(2-Chloroimidazo[1,2-a]pyridin-3-ylsulfonyl)-3-(4,6-dimethoxypyrimidin-2-yl)urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13ClN6O5S
CAS Number:
Molecular Weight:
412.81
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1cc(OC)nc(NC(=O)NS(=O)(=O)c2c(Cl)nc3ccccn23)n1

InChI

1S/C14H13ClN6O5S/c1-25-9-7-10(26-2)18-13(17-9)19-14(22)20-27(23,24)12-11(15)16-8-5-3-4-6-21(8)12/h3-7H,1-2H3,(H2,17,18,19,20,22)

InChI key

NAGRVUXEKKZNHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Imazosulfuron is known as a novel post-emergence herbicide that belongs to the sulfonylurea class of compounds. Its mode of action involves the regulation of annual and perennial broad-leaf weeds and sedges in rice products.[1]

Application

Imazosulfuron may be used as an analytical reference standard for the determination of the analyte in:
  • Water samples by liquid chromatography with electrospray ionization mass spectrometric detection (LC-ESI-MS).[2]
  • Paddy field water samples by direct injection using LC combined with hybrid triple-quadrupole-linear ion trap (QqLIT) MS.[3]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caitlin Rering et al.
Journal of agricultural and food chemistry, 65(15), 3103-3108 (2017-04-04)
Imazosulfuron, a sulfonylurea herbicide used in rice cultivation, has been shown to undergo photodegradation in water, but neither the photochemical mechanism nor the role of indirect photolysis is known. The purpose of this study was to investigate the underlying processes
Determination of imazosulfuron persistence in rice crop and soil.
Sondhia S.
Environmental Monitoring and Assessment, 137(1-3), 205-211 (2008)
Determination and quantitation of sulfonylurea and urea herbicides in water samples using liquid chromatography with electrospray ionization mass spectrometric detection.
Ayano E, et al.
Analytica Chimica Acta, 507(2), 211-218 (2004)
Trace analysis of pesticides in paddy field water by direct injection using liquid chromatography-quadrupole-linear ion trap-mass spectrometry
Pareja L, et al.
Journal of Chromatography A, 1218(30), 4790-4798 (2011)
Amin Mojiri et al.
Chemosphere, 253, 126646-126646 (2020-04-11)
Although pesticides are widely used in agriculture, industry and households, they pose a risk to human health and ecosystems. Based on target organisms, the main types of pesticides are herbicides, insecticides and fungicides, of which herbicides accounted for 46% of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service