Skip to Content
Merck
All Photos(1)

Key Documents

32489

Supelco

Cyclosporin A-13C62 solution

VETRANAL®, 25 μg/mL in acetonitrile, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C62H111N11O12
Molecular Weight:
1264.16
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

concentration

25 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

mass shift

M+62

storage temp.

−20°C

SMILES string

[13CH3][13CH2][13C@@H]1N[13C](=O)[13C@H]([13C@H](O)[13C@H]([13CH3])[13CH2]\[13CH]=[13CH]\[13CH3])N([13CH3])[13C](=O)[13C@H]([13CH]([13CH3])[13CH3])N([13CH3])[13C](=O)[13C@H]([13CH2][13CH]([13CH3])[13CH3])N([13CH3])[13C](=O)[13C@H]([13CH2][13CH]([13CH3])[13CH3])N([13CH3])[13C](=O)[13C@@H]([13CH3])N[13C](=O)[13C@H]([13CH3])N[13C](=O)[13C@H]([13CH2][13CH]([13CH3])[13CH3])N([13CH3])[13C](=O)[13C@@H](N[13C](=O)[13C@H]([13CH2][13CH]([13CH3])[13CH3])N([13CH3])[13C](=O)[13CH2]N([13CH3])[13C]1=O)[13CH]([13CH3])[13CH3]

InChI

1S/C62H111N11O12/c1-25-27-28-40(15)52(75)51-56(79)65-43(26-2)58(81)67(18)33-48(74)68(19)44(29-34(3)4)55(78)66-49(38(11)12)61(84)69(20)45(30-35(5)6)54(77)63-41(16)53(76)64-42(17)57(80)70(21)46(31-36(7)8)59(82)71(22)47(32-37(9)10)60(83)72(23)50(39(13)14)62(85)73(51)24/h25,27,34-47,49-52,75H,26,28-33H2,1-24H3,(H,63,77)(H,64,76)(H,65,79)(H,66,78)/b27-25+/t40-,41+,42-,43+,44+,45+,46+,47+,49+,50+,51+,52-/m1/s1/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1,15+1,16+1,17+1,18+1,19+1,20+1,21+1,22+1,23+1,24+1,25+1,26+1,27+1,28+1,29+1,30+1,31+1,32+1,33+1,34+1,35+1,36+1,37+1,38+1,39+1,40+1,41+1,42+1,43+1,44+1,45+1,46+1,47+1,48+1,49+1,50+1,51+1,52+1,53+1,54+1,55+1,56+1,57+1,58+1,59+1,60+1,61+1,62+1

InChI key

PMATZTZNYRCHOR-GCHSCARRSA-N

General description

Certan Vial

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service