Skip to Content
Merck
All Photos(1)

Documents

31458

Sigma-Aldrich

Magnesia grooves

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgO
Molecular Weight:
40.30
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

reaction suitability

reagent type: catalyst
core: magnesium

SMILES string

O=[Mg]

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

package of 100 pieces

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solaiman Tarafder et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 103(3), 679-690 (2014-07-22)
The presence of interconnected macro pores allows guided tissue regeneration in tissue engineering scaffolds. However, highly porous scaffolds suffer from having poor mechanical strength. Previously, we showed that microwave sintering could successfully be used to improve mechanical strength of macro
Lin Zhong et al.
Drug development and industrial pharmacy, 40(12), 1660-1669 (2013-10-09)
The amorphous solid dispersions of telmisartan salts were prepared by cogrinding, in presence of alkalizers and polyvinylpyrrolidone (PVPk30). Five alkalizers in this study were MgO, Na2CO3, K2CO3, NaHCO3 and meglumine. In soft mode using a roll mill, the drug could
Tobias C Keller et al.
ChemSusChem, 7(6), 1729-1738 (2014-04-24)
High-silica zeolites have been reported recently as efficient catalysts for liquid- and gas-phase condensation reactions because of the presence of a complementary source of basicity compared to Al-rich basic zeolites. Herein, we describe the controlled generation of these active sites
Ayuko Imaoka et al.
Drug metabolism and pharmacokinetics, 29(5), 414-418 (2014-05-09)
The drug interaction between new quinolone antibiotics (NQs) and polyvalent metal cation products, leading to a significant decrease in the absorption of NQ, is considered to be attributable to the formation of poorly absorbable chelate and physicochemical adsorption of NQs
Kazuhiro Iguchi et al.
Oncology letters, 7(5), 1665-1668 (2014-04-26)
Prostate cancer occurs more frequently among older males and such elderly individuals often have chronic underlying disorders for which various drugs are administered for treatment. The levels of prostate-specific antigen (PSA), a widely used prostate cancer marker, are influenced by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service