Skip to Content
Merck
All Photos(1)

Key Documents

31437-M

Sigma-Aldrich

Sodium bicarbonate

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.7%

Synonym(s):

Sodium hydrogen carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaHCO3
CAS Number:
Molecular Weight:
84.01
Beilstein:
4153970
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.7%
form:
powder
solubility:
H2O: 0.5 g/10 mL, clear, colorless
water: soluble 50 g/L

grade

ACS reagent
puriss. p.a.

Agency

reag. Ph. Eur.

Assay

≥99.7%

form

powder

impurities

≤0.003% S-compounds (as SO4)
≤0.015% in water insoluble matters
≤5 mg/kg ammonium (NH4)
≤50 mg/kg I red. matters (as SO2)

pH

≤8.6 (20 °C, 5%)

pKa (25 °C)

(1) 6.37, (2) 10.25 (carbonic acid)

solubility

H2O: 0.5 g/10 mL, clear, colorless
water: soluble 50 g/L

density

2.16 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.003%
phosphate (PO43-): ≤10 mg/kg

cation traces

As: ≤0.0002%
Ca: ≤0.01%
Cd: ≤0.0005%
Co: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.005%
Ni: ≤0.005%
Pb: ≤0.0005%
Zn: ≤0.0005%
heavy metals: ≤5 mg/kg (by ICP-OES)

suitability

complies for appearance of solution

SMILES string

[Na+].OC([O-])=O

InChI

1S/CH2O3.Na/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

UIIMBOGNXHQVGW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium bicarbonate is widely used as a physiological buffer. The pKa for bicarbonate ↔ carbonic acid reaction is 6.37. The pKa for bicarbonate ↔ carbonate reaction is 10.25.

Application

Sodium bicarbonate may be used to maintain 7.4pH in Iscove′s modified Dulbecco′s medium for growth of murine hybridoma cell culture.[1] It may be employed as a component of RPMI-1640 liquid medium in a study.[2]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D W Denning et al.
The Journal of antimicrobial chemotherapy, 40(3), 401-414 (1997-10-24)
Given the increased choice of therapeutic agents and the rising incidence of serious invasive disease, it is important that reliable in-vitro methods for detecting antifungal drug resistance in Aspergillus spp. are developed. Six clinical isolates of Aspergillus fumigatus, obtained from
S S Ozturk et al.
Biotechnology and bioengineering, 39(4), 418-431 (1992-02-20)
The influence of ammonia and lactate on cell growth, metabolic, and antibody production rates was investigated for murine hybridoma cell line 163.4G5.3 during batch culture. The specific growth rate was reduced by one-half in the presence of an initial ammonia

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service