Skip to Content
Merck
All Photos(1)

Key Documents

257400

Sigma-Aldrich

1,4-Bis(2-methylstyryl)benzene

≥99.5%

Synonym(s):

Bis-MSB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3C6H4CH=CH)2C6H4
CAS Number:
Molecular Weight:
310.43
Beilstein:
2375845
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99.5%

mp

180-182 °C (lit.)

λmax

349 nm

SMILES string

Cc1ccccc1\C=C\c2ccc(cc2)\C=C\c3ccccc3C

InChI

1S/C24H22/c1-19-7-3-5-9-23(19)17-15-21-11-13-22(14-12-21)16-18-24-10-6-4-8-20(24)2/h3-18H,1-2H3/b17-15+,18-16+

InChI key

QKLPIYTUUFFRLV-YTEMWHBBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable as laser dye

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service