Skip to Content
Merck
All Photos(1)

Documents

25460

Sigma-Aldrich

1-Chloro-2-nitrobenzene

purum, ≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NO2
CAS Number:
Molecular Weight:
157.55
Beilstein:
509273
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

5.4 (vs air)

vapor pressure

0.04 mmHg ( 25 °C)

grade

purum

Assay

≥99.0% (GC)

autoignition temp.

500 °F

expl. lim.

8.7 %

bp

246 °C (lit.)

mp

31-33 °C (lit.)

density

1.348 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1ccccc1Cl

InChI

1S/C6H4ClNO2/c7-5-3-1-2-4-6(5)8(9)10/h1-4H

InChI key

BFCFYVKQTRLZHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

258.8 °F - closed cup

Flash Point(C)

126 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoxuan Zhang et al.
Journal of hazardous materials, 177(1-3), 508-515 (2010-01-12)
Poly-3-hydroxybutyrate (PHB) was used as a new material to prepare a biomimetic adsorbent by a modified double emulsion solvent evaporation technique. The enrichment capacities of the adsorbent for the toxic liposoluble organic compounds were evaluated by chlorobenzene (CB) and o-nitrochlorobenzene
Hong Liu et al.
Biotechnology letters, 27(4), 275-278 (2005-03-03)
A strain of Pseudomonas stutzeri ZWLR2-1 was isolated from soil contaminated with chloronitrobenzenes and identified by 16S rDNA sequencing. This bacterium released chloride and nitrite into the medium when grown on 0.5 mM 2-chloronitrobenzene. PCR amplification and DNA sequencing revealed
Voltammetric determination of airborne 2-chloronitrobenzene using a recirculating absorbent vapour sampler.
P R Fielden et al.
The Analyst, 113(12), 1799-1802 (1988-12-01)
Michiharu Matsumoto et al.
The Journal of toxicological sciences, 31(1), 9-22 (2006-03-16)
Para- and ortho-chloronitrobenzene (p- and o-CNB) were compared for subchronic toxicity by feeding F344 rats and BDF(1) mice of both sexes p-CNB-or o-CNB-containing diets at 5 different concentrations for 13 weeks. The two isomers induced hematotoxicity and hepatotoxicity of different
G S Travlos et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 30(1), 75-92 (1996-03-01)
Toxicity studies were performed by exposing F344/N rats and B6C3F1 mice to 2- and 4-chloronitrobenzene (CNB) by whole-body inhalation 6 hr/day, 5 days/week, for 13 weeks. Animals were evaluated for clinical chemistry (rats), hematology (rats), histopathology, and body/organ weights. Exposure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service