Skip to Content
Merck
All Photos(1)

Key Documents

25035

Sigma-Aldrich

Chloromethylphosphonic dichloride

purum, ≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2P(O)Cl2
CAS Number:
Molecular Weight:
167.36
Beilstein:
969707
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

purum

Assay

≥97.0% (GC)

refractive index

n20/D 1.497 (lit.)
n20/D 1.497

bp

78-82 °C/10 mmHg (lit.)

density

1.638 g/mL at 25 °C (lit.)

SMILES string

ClCP(Cl)(Cl)=O

InChI

1S/CH2Cl3OP/c2-1-6(3,4)5/h1H2

InChI key

FZXSSJIKXCFPDP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service