Skip to Content
Merck
All Photos(1)

Key Documents

21327

Sigma-Aldrich

(1R,E)-(+)-Camphorquinone 3-oxime

purum, ≥97.0% (sum of enantiomers, HPLC)

Synonym(s):

(1R,E)-(+)-2,3-Bornanedione 3-oxime, anti-(1R)-(+)-Camphorquinone 3-oxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO2
CAS Number:
Molecular Weight:
181.23
Beilstein:
3200377
MDL number:
UNSPSC Code:
12352000

grade

purum

Assay

≥97.0% (sum of enantiomers, HPLC)

optical activity

[α]20/D +199±3°, c = 1% in ethanol

mp

153-156 °C (lit.)
153-156 °C

storage temp.

2-8°C

SMILES string

[H][C@@]12CC[C@@](C)(C(=O)\C1=N/O)C2(C)C

InChI

1S/C10H15NO2/c1-9(2)6-4-5-10(9,3)8(12)7(6)11-13/h6,13H,4-5H2,1-3H3/b11-7-/t6-,10+/m1/s1

InChI key

YRNPDSREMSMKIY-HAKKTOSXSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Chiral intermediate; starting material for the synthesis of cis-3-amino-borneol and -isoborneol, versatile chiral auxiliaries

comparable product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N. Ikota et al.
Chemical & Pharmaceutical Bulletin, 34, 1050-1050 (1986)
R.A. Chittenden et al.
Journal of the Chemical Society, 49-49 (1970)
Catalytic asymmetric induction. Highly enantioselective addition of dialkylzincs to aldehydes.
M Kitamura et al.
Journal of the American Chemical Society, 108(19), 6071-6072 (1986-09-01)
T.R. Kelly et al.
Tetrahedron Letters, 25, 39-39 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service