Skip to Content
Merck
All Photos(1)

Key Documents

210064

Sigma-Aldrich

Periodic acid

reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H5IO6
CAS Number:
Molecular Weight:
227.94
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

reagent grade

Assay

98%

reaction suitability

reagent type: oxidant

mp

122 °C (lit.)

SMILES string

OI(O)(O)(O)(O)=O

InChI

1S/H5IO6/c2-1(3,4,5,6)7/h(H5,2,3,4,5,6,7)

InChI key

TWLXDPFBEPBAQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 1 - Skin Corr. 1B - STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Murad Alturkustani et al.
Journal of neuropathology and experimental neurology, 74(3), 233-240 (2015-02-11)
The pathologic features of adult-onset leukoencephalopathy/leukodystrophy with axonal spheroids (ALAS) are variable, and this has led to different hypotheses as to whether primarily demyelination or axonopathy may underlie this disorder. Typical ALAS pathology is rarely accompanied by focal multiple sclerosis
Vered Padler-Karavani et al.
The Journal of biological chemistry, 287(27), 22593-22608 (2012-05-03)
DNA and protein arrays are commonly accepted as powerful exploratory tools in research. This has mainly been achieved by the establishment of proper guidelines for quality control, allowing cross-comparison between different array platforms. As a natural extension, glycan microarrays were
Karen Mollet et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(10), 3383-3396 (2013-01-26)
The reactivity of 3-hydroxy-4-(1,2-dihydroxyethyl)-β-lactams with regard to the oxidant sodium periodate was evaluated, unexpectedly resulting in the exclusive formation of new 2-hydroxy-1,4-oxazin-3-ones through a C3C4 bond cleavage of the intermediate 4-formyl-3-hydroxy-β-lactams followed by a ring expansion. This peculiar transformation stands
Dmitriy Berillo et al.
Macromolecular bioscience, 12(8), 1090-1099 (2012-06-08)
Macroporous scaffolds composed of chitosan and using oxidized dextran as a crosslinker are produced through cryogelation. Introducing gelatin as a third component into the structure results in the formation of mesopores in the pore walls, which are not seen if
D Canbaz et al.
Allergy, 70(5), 522-532 (2015-02-14)
The initial immune response to house dust mite (HDM) is orchestrated by an interplay between epithelial cells (ECs) and dendritic cells (DCs). Innate cytokines released by HDM-exposed ECs activate airway DCs and effector inflammatory cells, which together induce a HDM-specific

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service