Skip to Content
Merck
All Photos(1)

Key Documents

19750

Sigma-Aldrich

1-Chlorobutane

puriss. p.a., ≥99.5% (GC)

Synonym(s):

Butyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3Cl
CAS Number:
Molecular Weight:
92.57
Beilstein:
1730909
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

vapor density

3.2 (vs air)

vapor pressure

80.1 mmHg ( 78.4 °C)

grade

puriss. p.a.

Assay

≥99.5% (GC)

autoignition temp.

860 °F

expl. lim.

10.1 %

impurities

≤0.5% benzene (UV)

evapn. residue

≤0.001%

refractive index

n20/D 1.402 (lit.)
n20/D 1.402

bp

77-78 °C (lit.)

mp

−123 °C (lit.)

density

0.886 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CCCCCl

InChI

1S/C4H9Cl/c1-2-3-4-5/h2-4H2,1H3

InChI key

VFWCMGCRMGJXDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Lewandowicz et al.
Journal of the American Chemical Society, 123(19), 4550-4555 (2001-07-18)
We have found chlorine kinetic isotope effects on the dehalogenation catalyzed by haloalkane dehalogenase from Xanthobacter autotrophicus GJ10 to be 1.0045 +/- 0.0004 for 1,2-dichloroethane and 1.0066 +/- 0.0004 for 1-chlorobutane. The latter isotope effect approaches the intrinsic chlorine kinetic
J M Czuczwa et al.
Journal - Association of Official Analytical Chemists, 72(5), 752-759 (1989-09-01)
A gel permeation chromatographic (GPC) method, used by the U.S. Environmental Protection Agency (USEPA), was modified for cleanup of soil, sediments, wastes, and oily wastes before determination of semivolatile organic pollutants. The modifications included new calibration procedures and control of
M E Alburges et al.
Journal of analytical toxicology, 19(6), 381-386 (1995-10-01)
Ibogaine, an indolamine derivative, is currently being investigated as a potential agent in the treatment of stimulant and opiate addiction. We developed a rapid, sensitive, and specific method for the analysis of ibogaine and its putative active metabolite, 12-hydroxy-ibogamine (12-OH-ibogamine).
Boaz Shapira et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 182(1), 12-21 (2006-06-30)
We have recently proposed a protocol for retrieving nuclear magnetic resonance (NMR) spectra based on a spatially-dependent encoding of the MR interactions. It has also been shown that the spatial selectivity with which spins are manipulated during such encoding opens
Javier Catalán et al.
Physical chemistry chemical physics : PCCP, 13(33), 15022-15030 (2011-07-16)
In this work, we substantiate the change in the emitting state of indole caused by the dipolarity increase in the solvent 1-chlorobutane, observed on lowering the temperature from 293 to 133 K, accompanied by no significant changes in the corresponding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service