Skip to Content
Merck
All Photos(1)

Key Documents

17269

Supelco

Methyl all-cis-7,10,13,16,19-docosapentaenoate

analytical standard

Synonym(s):

cis-7,10,13,16,19-Docosapentaenoic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CHF 467.00

CHF 467.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
50 MG
CHF 467.00

About This Item

Empirical Formula (Hill Notation):
C23H36O2
CAS Number:
Molecular Weight:
344.53
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

CHF 467.00


Estimated to ship on30 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

functional group

ester

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCCCCC(=O)OC

InChI

1S/C23H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23(24)25-2/h4-5,7-8,10-11,13-14,16-17H,3,6,9,12,15,18-22H2,1-2H3/b5-4-,8-7-,11-10-,14-13-,17-16-

InChI key

PTFHIRHGARALFY-JEBPEJKESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl all-cis-7,10,13,16,19-docosapentaenoate is a fatty acid methyl ester.[1]

Application

It may be used as reference standard, for a study performed to investigate the effect of enzymatic transesterification of beeftallow, using experimental enzyme catalyst NS88001 with the short chain alcohol methanol and hexane.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Effectiveness of Enzymatic Transesterification of Beeftallow Using Experimental Enzyme Ns88001 with Methanol and Hexane.
Kumar, S., et al.
Enzyme Eng., 2.116, 1-10 (2013)
Robert Bodkowski et al.
Animals : an open access journal from MDPI, 10(10) (2020-10-15)
The study aimed to investigate the effect of supplementation of fish oil after the process of low-temperature crystallization (LTC-FO) enriched with long-chain polyunsaturated fatty acids (LC-PUFAs) on cow milk parameters. The experiment was carried out on 24 Polish Holstein Friesian

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service