Skip to Content
Merck
All Photos(1)

Key Documents

13425

Sigma-Aldrich

Sodium ferrocyanide decahydrate

≥98.0%

Synonym(s):

Tetrasodium hexacyanoferrate decahydrate, Sodium hexacyanoferrate(II)

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
CHF 71.20

CHF 71.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
CHF 71.20

About This Item

Linear Formula:
Na4Fe(CN)6 · 10H2O
CAS Number:
Molecular Weight:
484.06
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

CHF 71.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98.0%

form

solid

reaction suitability

core: iron
reagent type: catalyst

anion traces

chloride (Cl-): ≤0.20%
sulfate (SO42-): ≤0.20%

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].N#C[Fe-4](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/6CN.Fe.4Na.10H2O/c6*1-2;;;;;;;;;;;;;;;/h;;;;;;;;;;;10*1H2/q;;;;;;-4;4*+1;;;;;;;;;;

InChI key

KJSGVRTUMXJODL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sodium ferrocyanide decahydrate can be used as a precursor to synthesize Na-enriched Na1+xFe[Fe(CN)6] frameworks in NaCl, which can be used as a cathode for sodium-ion batteries.(1)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Francisco Jiménez-Fiérrez et al.
Sensors (Basel, Switzerland), 20(16) (2020-08-17)
Herein, a novel electrochemical glucose biosensor based on glucose oxidase (GOx) immobilized on a surface containing platinum nanoparticles (PtNPs) electrodeposited on poly(Azure A) (PAA) previously electropolymerized on activated screen-printed carbon electrodes (GOx-PtNPs-PAA-aSPCEs) is reported. The resulting electrochemical biosensor was validated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service