Skip to Content
Merck
All Photos(1)

Documents

134060

Saybolt Colour Reference Standard

ASTM D 6045, D 156 (ASTM colour 12)

Synonym(s):

Colour Reference Standard Saybolt 12, Saybolt Color Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

according to ASTM® D 156
according to ASTM® D 6045

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Saybolt colour (ASTM colour 12) reference standard is a certified reference material (CRM). This liquid colour reference standard is produced, packaged and stored following ISO 17034.

Please note that the value stated on this page is nominal. All certified values for the current lots of all Saybolt colour reference standards can be found on the Paragon Scientific webpage.

Application

Colour Reference Standard Saybolt 12 is used to calibrate colour measuring equipment such as colorimeters, employed in colour control experiments of process industry products.

It may be used as a colour standard in spectroscopic methods to test the quality of petroleum products.

Features and Benefits

  • Traceable to ISO 17025 / ISO 17034 under UKAS accreditation
  • Colour values in ASTM D 6045 and D 156 scales of measurement
  • 500-mL pack size available with safety data sheet
  • 18-months shelf life
  • Tamper-evident security packaging

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

replaced by

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service