Skip to Content
Merck
All Photos(2)

Key Documents

11220

Supelco

Brilliant Black BN

analytical standard

Synonym(s):

Food Black 1

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
CHF 197.00

CHF 197.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
CHF 197.00

About This Item

Empirical Formula (Hill Notation):
C28H17N5Na4O14S4
CAS Number:
Molecular Weight:
867.68
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
E Number:
E151
NACRES:
NA.24

CHF 197.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[Na+].[Na+].[Na+].CC(=O)Nc1ccc(c2cc(c(\N=N\c3ccc(\N=N/c4ccc(cc4)S([O-])(=O)=O)c5ccc(cc35)S([O-])(=O)=O)c(O)c12)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C28H21N5O14S4.4Na/c1-14(34)29-23-10-11-24(50(42,43)44)20-13-25(51(45,46)47)27(28(35)26(20)23)33-32-22-9-8-21(18-7-6-17(12-19(18)22)49(39,40)41)31-30-15-2-4-16(5-3-15)48(36,37)38;;;;/h2-13,35H,1H3,(H,29,34)(H,36,37,38)(H,39,40,41)(H,42,43,44)(H,45,46,47);;;;/q;4*+1/p-4/b31-30-,33-32+;;;;

InChI key

GMMAPXRGRVJYJY-OXYXOBGMSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Brilliant Black BN (BB) is a azo dye which is basically a tetra sodium salt. It is a synthetic dye mainly used as food additive. It is used as colouring agent to enhance the appearance. It is usually used in jellies, brown sauces and cake mixes.[1]

Application

It was used as colourant during quantification of synthetic colourants in fish roe and caviar for effective isolation using HPLC methods.[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew Joiner et al.
Journal of dentistry, 36 Suppl 1, S8-14 (2008-07-23)
To investigate a new optical approach to tooth whitening by enhancing the measurement and perception of tooth whiteness using blue coloured materials deposited onto the tooth surface. Salivary pellicle coated human extracted teeth or polished enamel specimens were used as
Liquid chromatographic quantification of synthetic colorants in fish roe and caviar.
Kirschbaum, Jochen, Corina Krause, and Hans Bruckner
European Food Research and Technology, 222.5-6, 572-579 (2006)
Violetta K Macioszek et al.
Cellular & molecular biology letters, 9(1), 107-122 (2004-03-30)
Additives, especially colors, are in widespread use in the food industry. With the exception of the quinolines, food colors are relatively weak mutagens and are certified as safe additives despite reports that some people have allergic reactions to them. The
Tao Meng et al.
Journal of virology, 93(17) (2019-06-07)
Hand, foot, and mouth disease (HFMD), a highly contagious disease in children, is caused by human enteroviruses, including enterovirus 71 (EV71), coxsackievirus A16 (CVA16), and coxsackievirus A6 (CVA6). Although HFMD is usually mild and self-limiting, EV71 infection occasionally leads to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service