09991
Zonyl® FSJ fluorosurfactant
~40% solids in water: isopropyl alcohol (3:1)
Synonym(s):
anionic phosphate surfactant
About This Item
Recommended Products
concentration
~40% solids in water: isopropyl alcohol (3:1)
density
1.12 g/mL±0.05 g/mL at 20 °C
Physical form
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service