Skip to Content
Merck
All Photos(1)

Key Documents

09954

Sigma-Aldrich

Propargyl p-toluenesulfonate

≥97.0% (GC)

Synonym(s):

Propargyl tosylate

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
CHF 234.00

CHF 234.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
25 ML
CHF 234.00

About This Item

Empirical Formula (Hill Notation):
C10H10O3S
CAS Number:
Molecular Weight:
210.25
Beilstein:
1912957
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CHF 234.00


Estimated to ship on29 May 2025


Request a Bulk Order

Assay

≥97.0% (GC)

refractive index

n20/D 1.530

density

1.215 g/mL at 20 °C (lit.)

functional group

tosylate

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1)S(=O)(=O)OCC#C

InChI

1S/C10H10O3S/c1-3-8-13-14(11,12)10-6-4-9(2)5-7-10/h1,4-7H,8H2,2H3

InChI key

LMBVCSFXFFROTA-UHFFFAOYSA-N

Related Categories

Application

Propargyl p-toluenesulfonate can be used as an initiator in the synthesis of linear and cyclic poly(2-isopropyl-2-oxazoline)s by cationic ring-opening polymerization of 2-isopropyl-2-oxazoline.[1]
It can also be used as a reagent to synthesize:
  • 2-hydroxy-4-pentynoic acid by an alkylation reaction with diethyl 2-acetamidomalonate followed by subsequent hydrolysis, decarboxylation, diazotization, and hydroxylation reactions.[2]
  • Furan derivatives by Pd-catalyzed reaction with acylchromates.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linear and cyclic poly (2-isopropyl-2-oxazoline) s for fine control of thermoresponsiveness
Jung Yongseok, et al.
European Polymer Journal, 88, 605-612 (2017)
An economical and safe procedure to synthesize 2-hydroxy-4-pentynoic acid: A precursor towards `clickable?biodegradable polylactide
Zhang Quanxuan, et al.
Beilstein Journal of Organic Chemistry, 10(1), 1365-1371 (2014)
Preparation of furans by palladium-catalyzed reaction of acylchromates and propargylic tosylates
Nakamura M, et al.
Heterocycles, 59(1), 333-345 (2003)
Lucca Trachsel et al.
ACS nano, 14(8), 10054-10067 (2020-07-07)
The physicochemical properties of cyclic polymer adsorbates are significantly influenced by the steric and conformational constraints introduced during their cyclization. These translate into a marked difference in interfacial properties between cyclic polymers and their linear counterparts when they are grafted
Yuji Odagaki et al.
Journal of pharmacological sciences, 125(2), 157-168 (2014-05-23)
The functional activation of Gi/o proteins coupled to muscarinic acetylcholine receptors (mAChRs) was investigated with the conventional guanosine-5'-O-(3-[(35)S]thio) triphosphate ([(35)S]GTPγS) binding assay in rat brain membranes. The most efficacious stimulation elicited by acetylcholine or carbachol (CCh) was obtained in striatal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service