09904
Zonyl® FSA fluorosurfactant
~25% solids in water: isopropyl alcohol (1:1)
Synonym(s):
anionic lithium carboxylate fluorosurfactant
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
anionic
concentration
~25% solids in water: isopropyl alcohol (1:1)
density
1.05 g/mL±0.05 g/mL at 20 °C
Legal Information
Zonyl is a registered trademark of E. I. du Pont de Nemours and Company
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
69.8 °F
Flash Point(C)
21 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chao Lu et al.
Analytical chemistry, 79(2), 666-672 (2007-01-16)
Gold nanoparticles (GNPs) capped with nonionic fluorosurfactant molecules (Zonyl FSN) were synthesized, and with the colloidal solution as a probe reagent, a new postcolumn colorimetric detection method for HPLC assay of homocysteine (Hcy) has been developed. The FSN-capped GNPs exhibited
V Caslavska et al.
Caries research, 25(1), 27-33 (1991-01-01)
After in vitro treatments of bovine enamel with NaF/Zonyl FSC and NH4F in-depth profiles of the chemical composition of the outermost zone of surface enamel were obtained with electron spectroscopy. The most important finding of the study was that after
Effects of laundering upon the removal of atrazine and metolachlor from cotton, cotton/polyester, and polyester fabrics treated with fluorochemical finishes.
A Shaw et al.
Bulletin of environmental contamination and toxicology, 66(3), 319-325 (2001-02-15)
V Caslavska et al.
Journal of dental research, 66(6), 1159-1161 (1987-06-01)
The purpose of this animal investigation was to determine whether the cariostatic effect of sodium fluoride can be enhanced by combining it with certain surfactants. The surfactants, Zonyl FSC and Lodyne S-110, were selected because they had been found to
Effect of abrasion on protective properties of polyester and cotton/polyester blend fabrics.
A Shaw et al.
Bulletin of environmental contamination and toxicology, 56(6), 935-941 (1996-06-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service