Skip to Content
Merck
All Photos(1)

Documents

09779

Sigma-Aldrich

Fmoc-12-Ado-OH

≥98.0% (TLC)

Synonym(s):

12-(Fmoc-amino)dodecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H35NO4
CAS Number:
Molecular Weight:
437.57
Beilstein:
9593537
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

≥98.0% (TLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

color

white

mp

114-118 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)CCCCCCCCCCCNC(=O)OCC1c2ccccc2-c3ccccc13

InChI

1S/C27H35NO4/c29-26(30)18-8-6-4-2-1-3-5-7-13-19-28-27(31)32-20-25-23-16-11-9-14-21(23)22-15-10-12-17-24(22)25/h9-12,14-17,25H,1-8,13,18-20H2,(H,28,31)(H,29,30)

InChI key

HVGIKYAQSSNFCH-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christos Kontos et al.
Nature communications, 11(1), 5981-5981 (2020-11-27)
Targeting a specific chemokine/receptor axis in atherosclerosis remains challenging. Soluble receptor-based strategies are not established for chemokine receptors due to their discontinuous architecture. Macrophage migration-inhibitory factor (MIF) is an atypical chemokine that promotes atherosclerosis through CXC-motif chemokine receptor-4 (CXCR4). However
Jing Han et al.
Molecular pharmaceutics, 15(7), 2840-2856 (2018-05-26)
GLP-1 analogs suffer from the main disadvantage of a short in vivo half-life. Lithocholic acid (LCA), one of the four main bile acids in the human body, possesses a high albumin binding rate. We therefore envisioned that a LCA-based peptide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service