Skip to Content
Merck
All Photos(1)

Key Documents

07566

Supelco

Difenoconazole-(4-chlorophenoxy-d4)

PESTANAL®, analytical standard

Synonym(s):

Difenoconazole-d4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19D4H13Cl2N3O3
Molecular Weight:
410.29
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥97.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

General description

Difenoconazole-(4-chlorophenoxy-d4) is an isotope-labeled analog of the broad-spectrum triazole fungicide difenoconazole, wherein 4-chlorophenoxy protons are substituted by deuterium.[1][2]

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiral triazole fungicide difenoconazole: absolute stereochemistry, stereoselective bioactivity, aquatic toxicity, and environmental behavior in vegetables and soil
Dong F, et al.
Environmental Science & Technology, 47(7), 3386-3394 (2013)
Residues and dissipation kinetics of triazole fungicides difenoconazole and propiconazole in wheat and soil in Chinese fields
Zhang Z, et al.
Food Chemistry, 168(7), 396-403 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service