Skip to Content
Merck
All Photos(1)

Key Documents

06927

Supelco

Monosodium N-lauroyl-L-glutamate

analytical standard

Synonym(s):

N-(1-Oxododecyl)-L-glutamic acid monosodium salt, N-Lauroyl-L-glutamic acid monosodium salt, Sodium N-dodecanoylglutamate, Sodium lauroyl glutamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H30NNaO5
CAS Number:
Molecular Weight:
351.41
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (TLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

SMILES string

OC(CC[C@H](NC(CCCCCCCCCCC)=O)C(O[Na])=O)=O

InChI

1S/C17H31NO5.Na/c1-2-3-4-5-6-7-8-9-10-11-15(19)18-14(17(22)23)12-13-16(20)21;/h14H,2-13H2,1H3,(H,18,19)(H,20,21)(H,22,23);/q;+1/p-1/t14-;/m0./s1

InChI key

IWIUXJGIDSGWDN-UQKRIMTDSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service