Skip to Content
Merck
All Photos(1)

Key Documents

06054

Sigma-Aldrich

L-Isoserine

≥98.0% (TLC)

Synonym(s):

(S)-2-Hydroxy-β-alanine, (S)-3-Amino-2-hydroxypropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7NO3
CAS Number:
Molecular Weight:
105.09
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (TLC)

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

color

white to off-white

mp

197-198 °C

application(s)

peptide synthesis

SMILES string

NC[C@H](O)C(O)=O

InChI

1S/C3H7NO3/c4-1-2(5)3(6)7/h2,5H,1,4H2,(H,6,7)/t2-/m0/s1

InChI key

BMYNFMYTOJXKLE-REOHCLBHSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaqin Sun et al.
Frontiers in pharmacology, 12, 628209-628209 (2021-03-13)
Promoting the differentiation of oligodendrocyte precursor cells (OPCs) is important for fostering remyelination in multiple sclerosis. Catalpol has the potential to promote remyelination and exert neuroprotective effects, but its specific mechanism is still unclear. Recent studies have shown that the
Jan Cz Dobrowolski et al.
Physical chemistry chemical physics : PCCP, 12(36), 10818-10830 (2010-07-10)
The IR low-temperature Ar and Kr matrix spectra of l-isoserine were registered for the first time and interpreted by means of the anharmonic DFT frequencies calculated at the B3LYP/aug-cc-pVTZ and B3LYP/aug-cc-pVDZ levels. 54 l-isoserine conformers were predicted to be stable
B S Coller et al.
The Journal of biological chemistry, 268(28), 20741-20743 (1993-10-05)
Peptides containing sequences derived from the new NH2 terminus of the seven-transmembrane domain thrombin receptor after thrombin cleavage can activate platelets directly. We recently demonstrated that such peptides are readily cleaved and inactivated by plasma, serum, and endothelial cell-associated aminopeptidase
Nagarjuna Palyam et al.
The Journal of organic chemistry, 74(11), 4390-4392 (2009-05-02)
We report a new protocol for synthesis of L-1-deoxymannojirimycin, L-1-deoxyidonojirimycin, and the N-isopropyl derivative of the latter compound from the readily available precursors (S)-isoserinal hydrate and 2-tert-butyl-2-methyl-1,3-dioxan-5-one. The key steps include diastereoselective proline-catalyzed syn aldol transformation and a reductive amination/cyclization.
Thomas Rühl et al.
Chemical communications (Cambridge, England), (15)(15), 1630-1631 (2002-08-13)
The photolytic decomposition of trifunctional carbene generating photoaffinity probes in methanolic solution was studied, a cleavage reaction with butylamine in water, the conjugation with a ligand (moenomycin), and experiments that demonstrate that the fully armed probes interact with penicillin-binding protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service