Skip to Content
Merck
All Photos(2)

Key Documents

05699

Supelco

Omeprazole sulfide 5-carboxylic acid

analytical standard

Synonym(s):

4-Methoxy-6-{[(5-methoxy-1H-benzimidazol-2-yl)thio]methyl}-5-methyl-3-pyridinecarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 1’540.00

CHF 1’540.00


Estimated to ship on17 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
CHF 1’540.00

About This Item

Empirical Formula (Hill Notation):
C17H17N3O4S
CAS Number:
Molecular Weight:
359.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

CHF 1’540.00


Estimated to ship on17 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1=C(CSC2=NC3=CC(OC)=CC=C3N2)N=CC(C(O)=O)=C1OC

InChI

1S/C17H17N3O4S/c1-9-14(18-7-11(16(21)22)15(9)24-3)8-25-17-19-12-5-4-10(23-2)6-13(12)20-17/h4-7H,8H2,1-3H3,(H,19,20)(H,21,22)

InChI key

BNOSHBGSJBEMGC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service