Skip to Content
Merck
All Photos(1)

Documents

05538

Millipore

Salmonella ChromoSelect Agar, Improved

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Salmonella Agar ChromoSelect, Improved

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 12.0 g/L
chromogenic mixture, 3.25 g/L
peptone special, 8.0 g/L
sodium deoxycholate, 1.0 g/L
yeast extract, 2.0 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.3±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages
water monitoring

microbiology

storage temp.

2-8°C

suitability

selective and differential for Escherichia coli
selective and differential for Salmonella spp.
selective and differential for coliforms

Related Categories

Application

A selective chromogenic medium used for simultaneous detection of Salmonella and Escherichia coli from food and water samples.

Preparation Note

Suspend 26.25 g in 1 litre distilled water. Boil gently to dissolve the medium completely. DO NOT AUTOCLAVE. Cool to 50°C. Mix well and pour into sterile plates.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service