Skip to Content
Merck
All Photos(1)

Key Documents

05433

Supelco

Conductivity Standard

conductivity 1413 μS/cm at 25 °C

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352302
PubChem Substance ID:

concentration

0.01 M KCl

density

1.00 g/mL at 20 °C

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deniz Ekinci et al.
Bioorganic & medicinal chemistry, 19(2), 744-748 (2011-01-08)
Carbonic anhydrase (CA; EC 4.2.1.1) was purified from the gill of the teleost fish Dicentrarchus labrax (European seabass). The purification procedure consisted of a single step affinity chromatography on Sepharose 4B-tyrosine-sulfanilamide. The enzyme was purified 84.9-fold with a yield of
Zhichao Liu et al.
PLoS computational biology, 7(12), e1002310-e1002310 (2011-12-24)
Drug-induced liver injury (DILI) is a significant concern in drug development due to the poor concordance between preclinical and clinical findings of liver toxicity. We hypothesized that the DILI types (hepatotoxic side effects) seen in the clinic can be translated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service