Skip to Content
Merck
All Photos(1)

Key Documents

05338

Sigma-Aldrich

1-Ethyl-2,3-dimethylimidazolium ethyl sulfate

BASF quality, ≥94.5% (HPLC)

Synonym(s):

Basionics® ST 67

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18N2O4S
CAS Number:
Molecular Weight:
250.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

BASF quality

Assay

≥94.5% (HPLC)

impurities

≤0.5% water

anion traces

chloride (Cl-): ≤100 mg/kg

SMILES string

CCOS([O-])(=O)=O.CCn1cc[n+](C)c1C

InChI

1S/C7H13N2.C2H6O4S/c1-4-9-6-5-8(3)7(9)2;1-2-6-7(3,4)5/h5-6H,4H2,1-3H3;2H2,1H3,(H,3,4,5)/q+1;/p-1

InChI key

OSCREXKVIJBLHA-UHFFFAOYSA-M

Legal Information

Basionics is a registered trademark of BASF SE

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

300.2 °F - closed cup

Flash Point(C)

149 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service