Skip to Content
Merck
All Photos(1)

Key Documents

04620590

(+)-Pulegone

primary reference standard

Synonym(s):

(R)-2-Isopropylidene-5-methylcyclohexanone, (R)-p-Menth-4(8)-en-3-one

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 283.00

CHF 283.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
CHF 283.00

About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
2040703
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

CHF 283.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

bp

223-224 °C (lit.)

density

0.935 g/mL at 20 °C (lit.)

application(s)

food and beverages

storage temp.

−20°C

SMILES string

C[C@@H]1CCC(=C(\C)C)\C(=O)C1

InChI

1S/C10H16O/c1-7(2)9-5-4-8(3)6-10(9)11/h8H,4-6H2,1-3H3/t8-/m1/s1

InChI key

NZGWDASTMWDZIW-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Other Notes

This compound is commonly found in plants of the genus: mentha

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service