Skip to Content
Merck
All Photos(1)

Key Documents

04007

Supelco

(1R)-(−)-Camphorquinone

analytical standard

Synonym(s):

(1R)-(−)-2,3-Bornanedione, 2,3-Bornanedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O2
CAS Number:
Molecular Weight:
166.22
Beilstein:
2327696
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

optical activity

[α]/D -106.0±4.0°, c = 0.2 in chloroform

shelf life

limited shelf life, expiry date on the label

mp

200-203 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

CC1(C)[C@@H]2CC[C@@]1(C)C(=O)C2=O

InChI

1S/C10H14O2/c1-9(2)6-4-5-10(9,3)8(12)7(6)11/h6H,4-5H2,1-3H3/t6-,10+/m1/s1

InChI key

VNQXSTWCDUXYEZ-LDWIPMOCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Camphorquinone, a 1,2-diketone, is a photoinitiator that finds wide use in the curing of resin composites. It functions by initiating the chain polymerization by free radical generation; typically with the aid of co-initiator amines.

Application

(1R)-(-)-Camphorquinone may be used as an analytical standard for the determination of the analyte in dental resin composite restorations in the oral environment in contact with food and beverages, and biological samples by analytical techniques.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

, book author=
Materials for the Direct Restoration of Teeth, 27(1), 195-212 (2016)
Sensitive chiral analysis by capillary electrophoresis
Garcia-Ruiz C and Marina ML
Electrophoresis, 27(1), 195-212 (2006)
Infrared spectroscopic analysis of restorative composite materials' surfaces and their saline extracts
Ajaj R, et al.
Progress in biomaterials, 2(1), 9-9 (2013)
D-erythrulose reductase can also reduce diacetyl: further purification and characterization of D-erythrulose reductase from chicken liver
Maeda M, et al.
Progress in biomaterials, 123(4), 602-606 (1998)
A visible light photochemical route to silver-epoxy nanocomposites by simultaneous polymerization-reduction approach
Yagci, Yusuf and Sangermano, Marco and Rizza, Giancarlo
Polymer, 49(24), 5195-5198 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service