Skip to Content
Merck
All Photos(1)

Documents

03906

Sigma-Aldrich

Ethylene oxide

purum, ≥99.8%

Synonym(s):

Oxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4O
CAS Number:
Molecular Weight:
44.05
Beilstein:
102378
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Assay

≥99.8%

refractive index

n20/D 1.3597 (lit.)

bp

10.7 °C (lit.)

mp

−111 °C (lit.)

density

0.882 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C1CO1

InChI

1S/C2H4O/c1-2-3-1/h1-2H2

InChI key

IAYPIBMASNFSPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Cylinder with net ~20 kg

Other Notes

Sales restrictions may apply

Recommended products

Z742162/Z742164 DIN 1 Regulators OR 99112 DIN 1 Valve

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Chem. Unst. Gas A - Eye Dam. 1 - Flam. Gas 1A - Muta. 1B - Press. Gas Liquefied gas - Repr. 1B - Skin Corr. 1B - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Nervous system, Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher J Whiteoak et al.
Inorganic chemistry, 51(20), 10639-10649 (2012-09-26)
Iron(III) amino triphenolate complexes with different substituents in the ortho-position of the phenolate moiety (R = H, Me, tBu, or Ph) have been synthesized by the reaction of iron(III) chloride and the sodium salt (Na(3)L(R)) of the requisite ligand. The
Faisal I Khan et al.
Journal of hazardous materials, 108(3), 147-159 (2004-05-04)
This paper discusses a methodology for the design of an optimum inspection and maintenance program. The methodology, called risk-based maintenance (RBM) is based on integrating a reliability approach and a risk assessment strategy to obtain an optimum maintenance schedule. First
Régis Coco et al.
International journal of pharmaceutics, 440(1), 3-12 (2012-07-24)
For inflammatory bowel disease (IBD) treatment, local delivery of molecules loaded in nanoparticles to the inflamed colon could be a promising strategy. The aim of this study was to investigate how drug-loaded polymeric nanoparticles target the site of inflammation and
V E Walker et al.
Mutation research, 233(1-2), 151-164 (1990-11-01)
The results of efforts to identify and quantify macromolecular adducts of ethylene oxide (ETO), to determine the source and significance of background levels of these adducts, and to generate molecular dosimetry data on these adducts are reviewed. A time-course study
Matthieu Gatineau et al.
American journal of veterinary research, 73(10), 1665-1669 (2012-09-28)
To compare effects of sterilization with hydrogen peroxide gas plasma (HPGP), ethylene oxide, and steam on bioadhesive properties of nylon and polyethylene lines used for stabilization of canine stifle joints. Samples of a 36.3-kg test nylon leader line, 57.8-kg test

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service