Skip to Content
Merck
All Photos(1)

Key Documents

03799

Supelco

Tungsten atomic spectroscopy standard concentrate 1.00 g W

1.00 g/L, for 1L standard solution, analytical standard

Synonym(s):

Sodium tungstate solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

packaging

ampule of

solubility

H2O: soluble

SMILES string

[Na+].[Na+].[O-][W]([O-])(=O)=O

InChI

1S/2Na.4O.W/q2*+1;;;2*-1;

InChI key

XMVONEAAOPAGAO-UHFFFAOYSA-N

Preparation Note

prepared with Na2WO4, NaOH and H2O

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Amigó-Correig et al.
PloS one, 7(7), e39087-e39087 (2012-07-18)
This study aims at exploring the effects of sodium tungstate treatment on hypothalamic plasticity, which is known to have an important role in the control of energy metabolism. Adult lean and high-fat diet-induced obese mice were orally treated with sodium
Josie E Bamford et al.
Environmental toxicology and chemistry, 30(10), 2312-2318 (2011-08-02)
Due to unknown effects of the potential exposure of the terrestrial environment to tungsten substances, a series of toxicity studies of sodium tungstate (Na(2) WO(4) ) was conducted. The effect on earthworm (Eisenia fetida) survival and reproduction was examined using
S M McInturf et al.
Toxicology and applied pharmacology, 254(2), 133-137 (2011-02-08)
The debate on tungsten (W) is fostered by its continuous usage in military munitions. Reports demonstrate W solubilizes in soil and can migrate into drinking water supplies and, therefore, is a potential health risk to humans. This study evaluated the
Laura Nocito et al.
PloS one, 7(8), e42305-e42305 (2012-08-21)
Oral administration of sodium tungstate has shown hyperglycemia-reducing activity in several animal models of diabetes. We present new insights into the mechanism of action of tungstate. We studied protein expression and phosphorylation in the liver of STZ rats, a type
C J Rodriguez-Hernandez et al.
FEBS letters, 586(3), 270-276 (2012-01-17)
Tungstate counteracts diabetes and obesity in animal models, but its molecular mechanisms remain elusive. Our Saccharomyces cerevisiae-based approach has found that tungstate alleviated the growth defect induced by nutrient stress and enhanced the activation of the GCN pathway. Tungstate relieved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service