Skip to Content
Merck
All Photos(1)

Documents

00840595

Vitexin

primary reference standard

Synonym(s):

4′,5,7-Trihydroxyflavone 8-C-glucoside, 8-C-Glucosylapigenin, Apigenin 8-C-glucoside, Orientoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O10
CAS Number:
Molecular Weight:
432.38
Beilstein:
67796
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)cc(O)c3C(=O)C=C(Oc23)c4ccc(O)cc4

InChI

1S/C21H20O10/c22-7-14-17(27)18(28)19(29)21(31-14)16-11(25)5-10(24)15-12(26)6-13(30-20(15)16)8-1-3-9(23)4-2-8/h1-6,14,17-19,21-25,27-29H,7H2/t14-,17-,18+,19-,21+/m1/s1

InChI key

SGEWCQFRYRRZDC-VPRICQMDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Other Notes

This compound is commonly found in plants of the genus: cannabis crataegus glycyrrhiza humulus

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tae-Gyu Nam et al.
Food chemistry, 170, 97-101 (2014-10-13)
It is known that common buckwheat sprouts contain several flavonoids, including orientin, isoorientin, vitexin, isovitexin, rutin, and quercetrin, whereas tartary buckwheat sprouts contain only rutin. In this study, we evaluated flavonoids present in buckwheat sprouts and identified a previously unreported
Hayoung Yoo et al.
Inflammation, 37(5), 1705-1717 (2014-04-29)
High mobility group box-1 (HMGB1) protein acts as a late mediator of severe vascular inflammatory conditions. Orientin has been known to have anxiolytic and antioxidative activities. However, the effect of orientin on HMGB1-induced inflammatory response has not been studied. We
Maryam Hamzeh-Mivehroud et al.
Archiv der Pharmazie, 347(10), 738-747 (2014-06-24)
Aldehyde oxidase (AO), an important enzyme in the biotransformation of drugs and xenobiotics, is inhibited by flavonoids. This enzyme can metabolize both aldehydes and N-heterocycles. In this work, a set of 15 flavonoids was assessed for inhibitory activity on the
Xin-shuai Wang et al.
Phytotherapy research : PTR, 29(3), 366-372 (2014-12-03)
In traditional therapy with Chinese medicine, vitexin has several pharmacological properties, including antinociceptive, antispasmodic, antioxidant, antimyeloperoxidase, and α-glucosidase inhibitory activities. Recently, vitexin was shown to protect the heart against ischemia/reperfusion injury in an in vitro model by inhibiting apoptosis. The
Clint M Alfaro et al.
Analytical chemistry, 86(21), 10639-10645 (2014-10-01)
With this study, we investigated why some small molecules demonstrate narrow dynamic ranges in electrospray ionization-mass spectrometry (ESI-MS) and sought to establish conditions under which the dynamic range could be extended. Working curves were compared for eight flavonoids and two

Protocols

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service