All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
grade:
SAJ special grade
form:
granular
Recommended Products
grade
SAJ special grade
vapor pressure
1 mmHg ( 487 °C)
form
granular
reaction suitability
reagent type: catalyst
core: zinc
availability
available only in Japan
resistivity
5.8 μΩ-cm, 20°C
bp
907 °C (lit.)
mp
420 °C (lit.)
density
7.133 g/mL at 25 °C (lit.)
suitability
suitable for arsenic determination
storage temp.
15-25°C
SMILES string
[Zn]
InChI
1S/Zn
InChI key
HCHKCACWOHOZIP-UHFFFAOYSA-N
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sara Eyal et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 50(5), 798-806 (2009-05-01)
Studies in rodents indicate that the disruption of P-glycoprotein (P-gp) function increases drug distribution into the developing fetus and organs such as the brain. To simultaneously and serially evaluate the effect of P-gp activity and inhibition on the tissue distribution
W Reainthippayasakul et al.
Journal of nanoscience and nanotechnology, 13(5), 3617-3621 (2013-07-19)
Conjugated meso-alkynyl 5,15-dimesitylporphyrin metal complexes have been synthesized by Sonogashira coupling reaction in good yields. Alkynyl groups were chosen as a link at the meso positions in order to extend the pi-conjugated length of porphyrin rings. These synthesized porphyrin derivatives
Bikha Ram Devrajani et al.
JPMA. The Journal of the Pakistan Medical Association, 63(3), 369-373 (2013-08-07)
To determine the serum zinc level in patients with pneumonia. The descriptive cross-sectional study was conducted at the Liaquat University Hospital, Hyderabad, Pakistan, from September 2008 to February 2009. All patients above 12 years of age and of either gender
Lena Martin et al.
Archives of animal nutrition, 67(3), 248-261 (2013-06-08)
This study aimed at investigating the effect of dietary zinc on performance, jejunal brush border membrane enzyme activities and mRNA levels of enzymes and two zinc transporters in piglets. A total of 126 piglets were weaned at 26 ±1 days
I A Popov et al.
Molekuliarnaia biologiia, 47(3), 498-504 (2013-07-31)
Alzheimer's disease, is a lethal neurodegenerative pathology, characterized by the formation of soluble neurotoxic oligomers of the human amyloid-beta peptide Abeta which get accumulated forming polymeric extracellular aggregates (so-called amyloid plaques). The isomerized at aspartate 7 isoform of the human
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service