Skip to Content
Merck
All Photos(1)

Documents

30-5530

Sigma-Aldrich

L-Tyrosine

SAJ special grade, ≥99.0%

Synonym(s):

(S)-2-Amino-3-(4-hydroxyphenyl)propionic acid, 3-(4-Hydroxyphenyl)-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
181.19
Beilstein:
392441
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

grade

SAJ special grade

Assay

≥99.0%

form

solid

availability

available only in Japan

mp

>300 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

N[C@@H](Cc1ccc(O)cc1)C(O)=O

InChI

1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m0/s1

InChI key

OUYCCCASQSFEME-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Amino acid precursor of dopamine and other catecholamines

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincent Dunet et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 53(2), 207-214 (2012-02-04)
For the past decade, PET with (18)F-fluoro-ethyl-tyrosine ((18)F-FET) has been used in the evaluation of patients with primary brain tumors (PBTs), but so far series have reported only a limited number of patients. The purpose of this systematic review and
Verena Rahm et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(4), 546-550 (2014-02-26)
Low-grade gliomas (LGGs) may harbor malignant foci, which are characterized by increased tumor cellularity and angiogenesis. We used diffusion-weighted MR imaging (apparent diffusion coefficient [ADC]) and PET with the amino acid O-(2-(18)F-fluorethyl)-L-tyrosine ((18)F-FET) to search for focal changes of diffusion
Keisuke Saito et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(19), 7690-7695 (2013-04-20)
Using quantum mechanics/molecular mechanics calculations and the 1.9-Å crystal structure of Photosystem II [Umena Y, Kawakami K, Shen J-R, Kamiya N (2011) Nature 473(7345):55-60], we investigated the H-bonding environment of the redox-active tyrosine D (TyrD) and obtained insights that help
Hideki Hara et al.
Nature immunology, 14(12), 1247-1255 (2013-11-05)
The inflammasome adaptor ASC contributes to innate immunity through the activation of caspase-1. Here we found that signaling pathways dependent on the kinases Syk and Jnk were required for the activation of caspase-1 via the ASC-dependent inflammasomes NLRP3 and AIM2.
T Eklundh et al.
Journal of psychiatric research, 30(3), 201-208 (1996-05-01)
In order to elucidate methodological aspects of CSF investigations, 14 healthy male subjects were lumbar-punctured at the L4-5 level following a standardised procedure. CSF concentrations of precursors, transmitters and transmitter metabolites were used as dependent variables, while age, height, body

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service