Skip to Content
Merck
All Photos(1)

Key Documents

30-3305

Sigma-Aldrich

Toluene

suitable for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
grade:
HPLC grade
technique(s):
HPLC: suitable
bp:
110-111 °C (lit.)
vapor pressure:
22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

grade

HPLC grade

vapor density

3.2 (vs air)

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

Assay

96.5-103.5% NaOH basis (titration)

form

liquid

autoignition temp.

997 °F

expl. lim.

7 %

availability

available only in Japan

technique(s)

HPLC: suitable

refractive index

n/D 1.496 (lit.)

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F

Flash Point(C)

4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Lei et al.
Small (Weinheim an der Bergstrasse, Germany), 11(24), 2946-2954 (2015-02-26)
Low-bandgap diketopyrrolopyrrole (DPP)-based polymers are used for the selective dispersion of semiconducting single-walled carbon nanotubes (s-SWCNTs). Through rational molecular design to tune the polymer-SWCNT interactions, highly selective dispersions of s-SWCNTs with diameters mainly around 1.5 nm are achieved. The influences
Liang Guo et al.
Journal of chromatography. A, 1286, 9-15 (2013-03-19)
For the first time, a novel one-step sample preparation method that combines microwave assisted extraction and solvent bar microextraction (MAE-SBME) with analysis by gas chromatography-mass spectrometry (GC-MS), was developed for the fast and efficient determination of polycyclic aromatic hydrocarbons (PAHs)
Elke Persch et al.
ChemMedChem, 9(8), 1880-1891 (2014-05-03)
The causative agents of the parasitic disease human African trypanosomiasis belong to the family of trypanosomatids. These parasitic protozoa exhibit a unique thiol redox metabolism that is based on the flavoenzyme trypanothione reductase (TR). TR was identified as a potential
Marina Shalaeva et al.
Journal of medicinal chemistry, 56(12), 4870-4879 (2013-05-29)
This study demonstrates that ΔlogP(oct-tol) (difference between logP(octanol) and logP(toluene)) describes compounds propensity to form intramolecular hydrogen bonds (IMHB) and may be considered a privileged molecular descriptor for use in drug discovery and for prediction of IMHB in drug candidates.
Tayseer Mahdi et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(31), 11134-11142 (2015-06-26)
Frustrated Lewis pairs (FLPs) based on sterically encumbered anilines and the Lewis acid B(C6 F5 )3 were found to react with terminal alkynes effecting intermolecular hydroamination affording iminium alkynylborate species of the form [RPhNC(R')Me][R'CCB(C6 F5 )3 ]. In these cases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service