Skip to Content
Merck
All Photos(1)

Key Documents

28-4630

Sigma-Aldrich

Sodium tripolyphosphate

SAJ first grade, 56.0-61.0% P2O5 basis

Synonym(s):

Pentasodium tripolyphosphate Anhydrous, STPP, Sodium triphosphate, Sodium triphosphate pentabasic, Sodium tripolyphosphate pentabasic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na5P3O10
CAS Number:
Molecular Weight:
367.86
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
SAJ first grade
form:
solid
Pricing and availability is not currently available.

grade

SAJ first grade

form

solid

availability

available only in Japan

concentration

56.0-61.0% P2O5

pH

9.5-10.3 (1 g/L)

storage temp.

15-25°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O

InChI

1S/5Na.H5O10P3/c;;;;;1-11(2,3)9-13(7,8)10-12(4,5)6/h;;;;;(H,7,8)(H2,1,2,3)(H2,4,5,6)/q5*+1;/p-5

InChI key

HWGNBUXHKFFFIH-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Fàbregas et al.
International journal of pharmaceutics, 446(1-2), 199-204 (2013-02-26)
Ionic gelation is the most frequently used method to obtain chitosan-tripolyphosphate nanoparticles due to its simplicity and because it does not generate waste solvents in the samples prepared. This paper presents a study of the physical factors involved in this
Michaela Weber et al.
Cell host & microbe, 13(3), 336-346 (2013-03-19)
Host defense to RNA viruses depends on rapid intracellular recognition of viral RNA by two cytoplasmic RNA helicases: RIG-I and MDA5. RNA transfection experiments indicate that RIG-I responds to naked double-stranded RNAs (dsRNAs) with a triphosphorylated 5' (5'ppp) terminus. However
Helene Jonassen et al.
Biomacromolecules, 13(11), 3747-3756 (2012-10-11)
The physical stability of chitosan nanoparticles cross-linked with sodium tripolyphosphate (TPP) was investigated over a period of 1 month. Special emphasis was placed on changes in the particle size and the particle compactness, which are two important physicochemical parameters of
Chih-Hui Yang et al.
Electrophoresis, 33(21), 3173-3180 (2012-09-06)
This study reports a facile method for the synthesis of hemispherical and discoidal chitosan microparticles by a combination of microfluidic technology and gelation strategy at an oil/water interface. Utilizing microfluidic emulsification in a cross-junction channel, the formation of regular droplets
Barbara Steigenberger et al.
Organic letters, 15(2), 366-369 (2013-01-05)
The synthesis of the triphosphates of 5-hydroxymethyl-, 5-formyl-, and 5-carboxycytidine and the incorporation of these building blocks into long DNA fragments using the polymerase chain reaction (PCR) are reported. In this way DNA fragments containing multiple hmC, fC, and caC

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service