Skip to Content
Merck
All Photos(1)

Key Documents

28-1190

Sigma-Aldrich

Silver chloride

SAJ special grade, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgCl
CAS Number:
Molecular Weight:
143.32
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥99.5%
grade:
SAJ special grade
form:
solid

grade

SAJ special grade

vapor pressure

1 mmHg ( 912 °C)

Assay

≥99.5%

form

solid

availability

available only in Japan

mp

455 °C (lit.)

storage temp.

15-25°C

SMILES string

Cl[Ag]

InChI

1S/Ag.ClH/h;1H/q+1;/p-1

InChI key

HKZLPVFGJNLROG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomohisa Hayashi et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 28(2), 127-133 (2012-02-11)
Electrochemical detection of sugar-related compounds was conducted using a boron-doped diamond (BDD) electrode as a detector for flow-injection analysis (FIA). Sugar-related compounds oxidize at high applied potentials, for which the BDD electrode is suitable for electrochemical measurements. Conditions for an
Alevtina Neyman et al.
Chemical communications (Cambridge, England), 48(16), 2207-2209 (2012-01-19)
"Out-of-pocket" association of Ag(+) to the tetradentate defect site of mono-vacant Keggin and Wells-Dawson polyoxometalate (POM) cluster-anions is used to direct the formation of water-soluble AgCl nanocubes.
Qiong Wang et al.
Biosensors & bioelectronics, 43, 19-24 (2013-01-01)
In this work, we describe a new sensitive strategy for electrochemical detection of protein via small molecule/protein interactions. This assay is based on a terminal protection mechanism that small molecule-linked single-stranded DNA (ssDNA) is protected against hydrolysis by exonuclease I
Siti Fauziyah Rahman et al.
Journal of microbiology and biotechnology, 22(10), 1446-1451 (2012-10-19)
Levodopa or L-3,4-dihydroxyphenylalanine (L-DOPA) is the precursor of the neurotransmitter dopamine. L-DOPA is a famous treatment for Parkinson's disease symptoms. In this study, electroenzymatic synthesis of L-DOPA was performed in a three-electrode cell, comprising a Ag/AgCl reference electrode, a platinum
Deliang Chen et al.
Nanoscale, 4(17), 5431-5439 (2012-07-28)
The hierarchical photocatalysts of Ag/AgCl@plate-WO₃ have been synthesized by anchoring Ag/AgCl nanocrystals on the surfaces of single-crystalline WO₃ nanoplates that were obtained via an intercalation and topochemical approach. The heterogeneous precipitation process of the PVP-Ag⁺-WO₃ suspensions with a Cl⁻ solution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service