Skip to Content
Merck
All Photos(1)

Documents

24-3440

Sigma-Aldrich

Chloroplatinic acid hexahydrate

JIS special grade, ≥98.5%

Synonym(s):

Hexachloroplatinic acid hexahydrate, Hydrogen hexachloroplatinate(IV) hexahydrate, Platinic chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2PtCl6 · 6H2O
CAS Number:
Molecular Weight:
517.90
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

JIS special grade

Assay

≥98.5%

form

solid

reaction suitability

reagent type: catalyst
core: platinum

availability

available only in Japan

SMILES string

O.O.O.O.O.O.Cl.Cl.Cl[Pt](Cl)(Cl)Cl

InChI

1S/6ClH.6H2O.Pt/h6*1H;6*1H2;/q;;;;;;;;;;;;+4/p-4

InChI key

PIJUVEPNGATXOD-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Han et al.
Scientific reports, 4, 6414-6414 (2014-09-19)
Noble metal nanoparticles with hollow interiors and customizable shell compositions have immense potential for catalysis. Herein, we present an unique structure transformation phenomenon for the fabrication of alloy Cu₃Pt nanoframes with polyhedral morphology. This strategy starts with the preparation of
Kevin M Marr et al.
ACS nano, 9(8), 7791-7803 (2015-06-25)
The utility of unmanned micro underwater vehicles (MUVs) is paramount for exploring confined spaces, but their spatial agility is often impaired when maneuvers require burst-propulsion. Herein we develop high-aspect ratio (150:1), multiwalled carbon nanotube microarray membranes (CNT-MMs) for propulsive, MUV
Sung Hoon Ahn et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 15(9), 1841-1848 (2014-05-13)
A facile way of controlling the structure of TiO(2) by changing the amount of water to improve the efficiency of dye-sensitized solar cells (DSSCs) is reported. Hierarchically ordered TiO(2) films with high porosity and good interconnectivity are synthesized in a
Chong Xiao et al.
The Analyst, 140(11), 3753-3758 (2015-01-30)
Here, we report a self-supported nanoporous gold microelectrode decorated with well-dispersed and tiny platinum nanoparticles as an electrochemical nonenzymatic hydrogen peroxide biosensor. Nanoporous gold was fabricated by electrochemical alloying/dealloying and then small-sized platinum nanoparticles were electrodeposited uniformly on them. This
Rui-Xiang Wang et al.
Nanoscale, 6(24), 14999-15007 (2014-11-05)
A novel nanostructured catalyst of platinum nanoparticles supported on 5,10,15,20-tetrakis(1-methyl-4-pyridinio)porphyrin tetra(p-toluenesulfonate) (TMPyP) functionalized graphene (TMPyP-graphene) is synthesized by the hydrothermal polyol process. The as-synthesized nanocomposites are characterized by Fourier transform infrared (FTIR) spectroscopy, UV-vis absorption spectroscopy, Raman spectroscopy, X-ray diffraction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service