Skip to Content
Merck
All Photos(1)

Key Documents

15-1520

Sigma-Aldrich

Iron(II) sulfide

SAJ first grade, ≥50.0%

Synonym(s):

Ferrous sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
FeS
CAS Number:
Molecular Weight:
87.91
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
SAJ first grade
form:
solid
Pricing and availability is not currently available.

grade

SAJ first grade

form

solid

reaction suitability

reagent type: catalyst
core: iron

availability

available only in Japan

concentration

≥50.0%

density

4.84 g/mL at 25 °C (lit.)

SMILES string

S=[Fe]

InChI

1S/Fe.S

InChI key

MBMLMWLHJBBADN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elvira Bura-Nakić et al.
Environmental science & technology, 47(2), 741-749 (2012-12-18)
Recent publications have shown that the anodic reaction between FeS and Hg can be used for electrochemical detection of colloidal and particulate FeS in natural waters. Anodic waves that were recorded around -0.45 V (vs Ag/AgCl) in model solutions correspond
Eun-Ju Kim et al.
ACS applied materials & interfaces, 3(5), 1457-1462 (2011-04-28)
Multicomponent nanoparticles containing two or more different types of functionalities show unique physical and chemical properties, leading to significantly enhanced performance. In this study, we have developed a new one-pot method to prepare Fe/FeS nanoparticles using dithionite at room temperature.
Insights into the mechanistic role of the [Fe4S4] cubane in the A-cluster {[Fe4S4]-(SR)-[NipNid]} of acetyl-coenzyme A synthase.
Yi Liu et al.
Chembiochem : a European journal of chemical biology, 12(9), 1417-1421 (2011-06-01)
Bo Zhang et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(39), 15734-15739 (2012-09-29)
Fumarate and nitrate reduction (FNR) regulatory proteins are O(2)-sensing bacterial transcription factors that control the switch between aerobic and anaerobic metabolism. Under anaerobic conditions [4Fe-4S](2+)-FNR exists as a DNA-binding homodimer. In response to elevated oxygen levels, the [4Fe-4S](2+) cluster undergoes
Bin Huang et al.
Bioresource technology, 102(21), 10154-10157 (2011-09-07)
The performance of acetic acid-supported pH-heterogenized heterotrophic denitrification (HD) facilitated with ferrous sulfide-based autotrophic denitrification (AD) was investigated in upflow activated carbon-packed column reactors for reliable removal of highly elevated nitrate (42 mg NO(3)-Nl(-1)) in drinking water. The use of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service