Skip to Content
Merck
All Photos(1)

Key Documents

07-4150

Sigma-Aldrich

p-Dimethylaminobenzylidenerhodanine

JIS special grade

Synonym(s):

5-(4-Dimethylaminobenzylidene)rhodanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2OS2
CAS Number:
Molecular Weight:
264.37
Beilstein:
189065
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

grade

JIS special grade

form

crystalline

availability

available only in Japan

mp

285-288 °C (lit.)

SMILES string

CN(C)c1ccc(cc1)\C=C2\SC(=S)NC2=O

InChI

1S/C12H12N2OS2/c1-14(2)9-5-3-8(4-6-9)7-10-11(15)13-12(16)17-10/h3-7H,1-2H3,(H,13,15,16)/b10-7+

InChI key

JJRVRELEASDUMY-JXMROGBWSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J W Ludlow et al.
Analytical biochemistry, 154(1), 104-109 (1986-04-01)
5-(4-Dimethylaminobenzylidene)rhodamine, a silver-specific dye, was used in a colorimetric assay to quantitate the autoradiographic deposition of silver onto X-ray film after exposure to sodium dodecyl sulfate-polyacrylamide gels of radiolabeled biological material. Silver grains were eluted from autoradiograms with 5 N
P Emanuele et al.
Annals of diagnostic pathology, 2(2), 125-126 (1998-12-10)
We report a modified copper stain that can be completed in 2 hours, rather than overnight, which is required for other stains. This is useful in the diagnosis of chronic cholestatic syndromes, Wilson's disease, Indian childhood cirrhosis, and other conditions
J S Kumaratilake et al.
Research in veterinary science, 42(1), 73-81 (1987-01-01)
Six Merino sheep were dosed orally with a 0.2 per cent solution of copper sulphate, six others were undosed controls. Liver biopsies were obtained and stained for copper by the p-dimethylaminobenzylidene rhodanine (DMABR), rubeanic acid (RA) and ferricyanide (FCN) methods

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service