Skip to Content
Merck
All Photos(1)

Key Documents

05-7490

Sigma-Aldrich

L-Cystine

SAJ special grade, ≥99.0%

Synonym(s):

(R,R)-3,3′-Dithiobis(2-aminopropionicacid)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-SCH2CH(NH2)CO2H]2
CAS Number:
Molecular Weight:
240.30
Beilstein:
1728094
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

grade

SAJ special grade

Assay

≥99.0%

availability

available only in Japan

mp

>240 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O

InChI

1S/C6H12N2O4S2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)/t3-,4-/m0/s1

InChI key

LEVWYRKDKASIDU-IMJSIDKUSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serge-George Rosolen et al.
Advances in experimental medicine and biology, 775, 145-154 (2013-02-09)
In the 70s, the amino acid taurine was found essential for photoreceptor survival. Recently, we found that taurine depletion can also trigger retinal ganglion cell degeneration both in vitro and in vivo. Therefore, evaluation of taurine levels could be a
Philip J Hogg
Nature reviews. Cancer, 13(6), 425-431 (2013-05-11)
Protein action in nature is generally controlled by the amount of protein produced and by chemical modification of the protein, and both are often perturbed in cancer. The amino acid side chains and the peptide and disulphide bonds that bind
Martijn J Wilmer et al.
American journal of physiology. Renal physiology, 299(5), F905-F916 (2010-09-10)
Renal proximal tubules are highly sensitive to ischemic and toxic insults and are affected in diverse genetic disorders, of which nephropathic cystinosis is the most common. The disease is caused by mutations in the CTNS gene, encoding the lysosomal cystine
Anne C Conibear et al.
The Journal of biological chemistry, 288(15), 10830-10840 (2013-02-23)
θ-Defensins are ribosomally synthesized cyclic peptides found in the leukocytes of some primate species and have promising applications as antimicrobial agents and scaffolds for peptide drugs. The cyclic cystine ladder motif, comprising a cyclic peptide backbone and three parallel disulfide
Maisie Lo et al.
Journal of cellular physiology, 215(3), 593-602 (2008-01-09)
The x(c) (-) cystine/glutamate antiporter is a major plasma membrane transporter for the cellular uptake of cystine in exchange for intracellular glutamate. Its main functions in the body are mediation of cellular cystine uptake for synthesis of glutathione essential for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service