Skip to Content
Merck
All Photos(1)

Key Documents

05-2745

Sigma-Aldrich

Chloride standard solution

1 mg/mL Cl-

Synonym(s):

Sodium chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

form

liquid

availability

available only in Japan

concentration

1 mg/mL Cl-

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francis Kim et al.
JAMA, 311(1), 45-52 (2013-11-19)
Hospital cooling improves outcome after cardiac arrest, but prehospital cooling immediately after return of spontaneous circulation may result in better outcomes. To determine whether prehospital cooling improves outcomes after resuscitation from cardiac arrest in patients with ventricular fibrillation (VF) and
C M Smart et al.
Anaesthesia, 69(2), 111-117 (2014-01-05)
To maximise the effect of a small fluid load, it is occasionally desirable to bolus manually with multiple depressions of a large-capacity syringe. This is usually achieved by placing the syringe on the side port of a three-way tap. We
Jessica J Buchanan et al.
Environmental technology, 34(5-8), 695-701 (2013-07-11)
Coal seam gas (CSG) is being touted as a transition fuel as the world moves towards low-carbon economies. However, the development of CSG reserves will generate enormous volumes of saline water. In this work, we investigate the potential of using
Jelle W G van den Berg et al.
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery, 150(1), 16-21 (2013-11-19)
To assess the effectiveness of nasal saline irrigation in adult patients with chronic rhinosinusitis. PubMed, EMBASE, the Cochrane Library. A comprehensive search was performed, and 2 authors independently screened publications. The design of selected studies was assessed on directness of
Yali V Zhang et al.
Science (New York, N.Y.), 340(6138), 1334-1338 (2013-06-15)
Below a certain level, table salt (NaCl) is beneficial for animals, whereas excessive salt is harmful. However, it remains unclear how low- and high-salt taste perceptions are differentially encoded. We identified a salt-taste coding mechanism in Drosophila melanogaster. Flies use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service